missing translation for 'onlineSavingsMsg'
Få mere at vide

(2E,4E)-2,4-Tetradecadienoic acid, TRC

Artikelnummer. 30418336
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418336 25 mg
30424055 50 mg
30416107 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30418336 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29240025MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 24738-49-6
Molekylær formel C14 H24 O2
Molekylvægt (g/mol) 224.34
Synonym (2E,4E)-2,4-Tetradecadienoic acid, (2E,4E)-Tetradecadienoic acid, (E,E)-Tetradeca-2,4-dienoic acid
IUPAC navn (2E,4E)-tetradeca-2,4-dienoic acid
SMIL CCCCCCCCC\C=C\C=C\C(=O)O

Tekniske data

Kemisk navn eller materiale (2E,4E)-2,4-Tetradecadienoic acid
Mængde 25 mg
InChI formel InChI=1S/C14H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h10-13H,2-9H2,1H3,(H,15,16)/b11-10+,13-12+
Formel vægt 224.1776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.