missing translation for 'onlineSavingsMsg'
Få mere at vide

(2E)-3-(Phenyl-d5)-2-propenoic Acid, TRC

Artikelnummer. 30427833
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427833 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427833 Leverandør Toronto Research Chemicals Leverandørnr. TRCP33618650MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2E)-3-(Phenyl-d5)-2-propenoic Acid
CAS 352431-48-2
Molekylær formel C9 D5 H3 O2
Mængde 50 mg
Synonym 2-Propenoic acid','3-(phenyl-d5)-','(2E)- (9CI), (2E)-3-(Phenyl-2,3,4,5,6-d5)-2-propenoic acid, trans-Cinnamic-d5 acid, (2E)-3-(Phenyl-d5)-2-propenoic acid, trans-Cinnamic acid D5 (phenyl-D5)
InChI formel InChI=1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(\C=C\C(=O)O)c([2H])c1[2H]
IUPAC navn (E)-3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoic acid
Molekylvægt (g/mol) 153.189
Formel vægt 153.084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.