Få mere at vide
(2E)-3-Methyl-2-hexenoic Acid, TRC
Artikelnummer. 30472615
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472615 50 mg
30471470 25 mg
30482383 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30472615 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33614050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27960-21-0
Molekylær formel C7H12O2
Molekylvægt (g/mol) 128.17
Synonym (E)-3-Methyl-2-hexenoic Acid, trans-3-Methyl-2-hexenoic Acid
IUPAC navn (E)-3-methylhex-2-enoic acid
SMIL CCC\C(=C\C(=O)O)\C
Tekniske data
Kemisk navn eller materiale (2E)-3-Methyl-2-hexenoic Acid
Mængde 50 mg
InChI formel InChI=1S/C7H12O2/c1-3-4-6(2)5-7(8)9/h5H,3-4H2,1-2H3,(H,8,9)/b6-5+
Formel vægt 128.0837