Learn More
(2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride, Mikromol™
Product Code. 30049176
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30049176 25 mg 25mg
1 options
Product Code. 30049176 Supplier Mikromol Supplier No. MM3376.030025
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride
CAS 661489-23-2
Analyte or Component Names (2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H12 N2 . Cl H
Certifications/Compliance ISO 9001
Synonym 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, hydrochloride (1:1), (2E)-, 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, monohydrochloride, (2E)- (9CI), (E)-3-(4-Amino-3,5-dimethylphenyl)-2-propenenitrile hydrochloride
InChI Formula InChI=1S/C11H12N2.ClH/c1-8-6-10(4-3-5-12)7-9(2)11(8)13;/h3-4,6-7H,13H2,1-2H3;1H/b4-3+;
SMILES Cl.Cc1cc(\C=C\C#N)cc(C)c1N
IUPAC Name (E)-3-(4-amino-3,5-dimethylphenyl)prop-2-enenitrile;hydrochloride
Molecular Weight (g/mol) 208.69
Quantity 25 mg
Formula Weight 208.0767 g/mol