Få mere at vide
(2E)-2,4-Pentadienoic Acid, TRC
Artikelnummer. 30426014
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426014 500 mg
30412567 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30426014 Leverandør Toronto Research Chemicals Leverandørnr. TRCP268350500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2E)-2,4-Pentadienoic Acid
CAS 21651-12-7
Molekylær formel C5H6O2
Mængde 500 mg
Synonym (E)-1,3-Butadiene-1-carboxylic Acid, trans-1-Carboxybutadiene, trans-2,4-Pentadienoic Acid
InChI formel InChI=1S/C5H6O2/c1-2-3-4-5(6)7/h2-4H,1H2,(H,6,7)/b4-3+
SMIL OC(=O)\C=C\C=C
IUPAC navn (2E)-penta-2,4-dienoic acid
Molekylvægt (g/mol) 98.1
Formel vægt 98.0368