Få mere at vide
(2beta,3alpha,5alpha,16beta,17beta)-2-(4-Morpholinyl)-16-(1-pyrrolidinyl)androstane-3,17-diol, TRC
Artikelnummer. 30135613
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135613 1 g
30009075 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135613 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7251451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 119302-20-4
Molekylær formel C27 H46 N2 O3
Molekylvægt (g/mol) 446.67
Synonym Androstane-3,17-diol, 2-(4-morpholinyl)-16-(1-pyrrolidinyl)-, (2β,3α,5α,16β,17β)-, (2β,3α,5α,16β,17β)-2-(4-Morpholinyl)-16-(1-pyrrolidinyl)androstane-3,17-diol, (2β,3α,5α,16β,17β)-2-(4-Morpholinyl)-16-(1-pyrrolidinyl)androstan-3,17-diol, 2-(4-Morpholinyl)-16-(pyrrolidin-1-yl)androstane-3,17-diol, 2β-(4-Morpholinyl)-16β-(1-pyrrolidinyl)-5α-androstane-3α,17β-diol, 2β-(Morpholin-4-yl)-16β-(pyrrolidin-1-yl)-5α-androstane-3α,17β-diol
IUPAC navn (2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-10,13-dimethyl-2-morpholin-4-yl-16-pyrrolidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
SMIL C[C@]12C[C@@H]([C@@H](O)C[C@@H]1CC[C@@H]3[C@@H]2CC[C@]4(C)[C@@H](O)[C@H](C[C@@H]34)N5CCCC5)N6CCOCC6
Tekniske data
Kemisk navn eller materiale (2beta,3alpha,5alpha,16beta,17beta)-2-(4-Morpholinyl)-16-(1-pyrrolidinyl)androstane-3,17-diol
Mængde 1 g
InChI formel InChI=1S/C27H46N2O3/c1-26-8-7-20-19(21(26)16-22(25(26)31)28-9-3-4-10-28)6-5-18-15-24(30)23(17-27(18,20)2)29-11-13-32-14-12-29/h18-25,30-31H,3-17H2,1-2H3/t18-,19+,20-,21-,22-,23-,24-,25-,26-,27-/m0/s1
Formel vægt 446.3508