missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human HIF-2 alpha/EPAS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2886-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot.
Tekniske data
| HIF-2 alpha/EPAS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99814 | |
| EPAS1 | |
| E. coli-derived recombinant human HIF-2 alpha/EPAS1 Ser543-Thr870 Accession # Q99814 | |
| 25 μg | |
| Primary | |
| Detects human HIF-2 alpha/EPAS1 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 318404 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2034 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion