missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Collagen V alpha 2 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00001290-Q01-25ug
Additional Details : Weight : 0.00970kg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification | |
1290 | |
25 μg | |
AB collagen, collagen alpha-2(V) chain, collagen, fetal membrane, A polypeptide, collagen, type V, alpha 2, MGC105115, type V preprocollagen alpha 2 chain | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
M.W. (Theoretical): 34.98 kDa | |
Store at -80C. Avoid freeze-thaw cycles. | |
COL5A2 | |
Collagen V alpha 2 |