Få mere at vide
(25R)-5Beta-Cholestane-3Alpha,7Alpha,12Alpha,26-tetrol, TRC
Artikelnummer. 30362427
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362427 10 mg
30354569 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362427 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43177510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27857-14-3
Molekylær formel C27H48O4
Molekylvægt (g/mol) 436.68
Synonym (3alpha,5beta,7alpha,12alpha,25R)-Cholestane-3,7,12,26-tetrol
IUPAC navn (3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-17-[(2R,6R)-7-hydroxy-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,7,12-triol
SMIL C[C@@H](CO)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Tekniske data
Kemisk navn eller materiale (25R)-5beta-Cholestane-3alpha,7alpha,12alpha,26-tetrol
Mængde 10 mg
InChI formel InChI=1S/C27H48O4/c1-16(15-28)6-5-7-17(2)20-8-9-21-25-22(14-24(31)27(20,21)4)26(3)11-10-19(29)12-18(26)13-23(25)30/h16-25,28-31H,5-15H2,1-4H3/t16-,17-,18+,19-,20-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formel vægt 436.3553