Learn More
Abnova™ 25 mM MgCl2
Product Code. 16092565
Change view
Click to view available options
Quantity:
1.8 mL
5 x 1.8 mL
Unit Size:
1.8mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16092565 5 x 1.8 mL Pack of 5
16082565 1.8 mL 1.8mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16092565 Supplier Abnova™ Supplier No. D0023.5x1.8mL
 Restricted Item
Forventet forsendelse: 27-01-2027
25 mM MgCl2 can be used for optimization of magnesium ion concentration in PCR reaction.
Specifications
For Use With (Application) Standard PCR
Quantity 5 x 1.8 mL