Få mere at vide
Tocris Bioscience™ 25-Hydroxycholesterol
GREENER_CHOICE
Antiviral; also α-crystallin (cryAB) chaperone
Brand:  Tocris Bioscience™ 5741/10
Encompass
Produktkode 15716765
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
25-Hydroxycholesterol is an antiviral. Stimulates expression of miR-185 microRNA which inhibits the hepatitis C virus life cycle via inhibition of cholesterol biosynthesis and virus cell membrane fusion. Also α-crystallin (cryAB) chaperone (Kd = 10.1 μM). Blocks α-crystallin aggregation and partially reverses cataract formation by binding and stabilizing soluble forms of cryAB in vitro and in vivo.
Specifications
Specifications
Cholest-5-ene-3β,25-diol
2140-46-7
402.65
Antiviral Compounds
Antiviral, Crystalline (cryAB) Chaperone
C27H46O2
Store at -20°C
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only