Få mere at vide
25-Hydroxy Vitamin D3 3,3’-Aminopropyl Ether, TRC
Artikelnummer. 30133188
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133188 25 mg -
30009842 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133188 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99583525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163018-26-6
Molekylær formel C30 H51 N O2
Molekylvægt (g/mol) 457.73
Synonym (εR,1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-(3-Aminopropoxy)-2-methylenecyclohexylidene]ethylidene]octahydro-α,α,ε,7a-tetramethyl-1H-indene-1-pentanol, (3β,5Z,7E)-3-(3-Aminopropoxy)-9,10-secocholesta-5,7,10(19)-trien-25-ol, 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3;
IUPAC navn (6R)-6-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-(2-aminoethoxy)-2-methylidenecyclohexylidene]ethylidene]-3a,7a-dimethyl-1,2,3,5,6,7-hexahydroinden-1-yl]-2-methylheptan-2-ol
SMIL C[C@H](CCCC(C)(C)O)[C@H]1CC[C@@]2(C)\C(=C\C=C/3\C[C@H](CCC3=C)OCCN)\CCC[C@]12C
Tekniske data
Kemisk navn eller materiale 25-Hydroxy Vitamin D3 3,3'-Aminopropyl Ether
Mængde 25 mg
InChI formel InChI=1S/C30H51NO2/c1-22-11-14-26(33-20-19-31)21-24(22)12-13-25-10-8-17-30(6)27(15-18-29(25,30)5)23(2)9-7-16-28(3,4)32/h12-13,23,26-27,32H,1,7-11,14-21,31H2,2-6H3/b24-12-,25-13+/t23-,26+,27-,29+,30-/m1/s1
Formel vægt 457.392