Få mere at vide
25-Hydroxy Vitamin D2 3,3’-Aminopropyl Ether (>90%), TRC
Artikelnummer. 30404064
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30404064 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404064 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9958251MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 25-Hydroxy Vitamin D2 3,3'-Aminopropyl Ether
CAS 932023-11-5
Molekylær formel C31H51NO2
Mængde 1 mg
Synonym (3S,4E,6R)-6-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-(3-Aminopropoxy)-2-methylenecyclohexylidene]ethylidene]octahydro-7a-methyl-1H-inden-1-yl]-2,3-dimethyl-4-hepten-2-ol
InChI formel InChI=1S/C31H51NO2/c1-22-11-15-27(34-20-8-19-32)21-26(22)14-13-25-9-7-18-31(6)28(16-17-29(25)31)23(2)10-12-24(3)30(4,5)33/h10,12-14,23-24,27-29,33H,1,7-9,11,15-21,32H2,2-6H3/b12-10+,25-13+,26-14-/t23-,24+,27+,28-,29+,31-/m1/s1
SMIL C[C@H](/C=C/[C@H](C)C(C)(C)O)[C@H]1CC[C@@H]\2[C@@]1(CCC/C2=C\C=C/3\C[C@H](CCC3=C)OCCCN)C
IUPAC navn (3S,6R,E)-6-((1R,3aS,7aR,E)-4-((Z)-2-((S)-5-(3-aminopropoxy)-2-methylenecyclohexylidene)ethylidene)-7a-methyloctahydro-1H-inden-1-yl)-2,3-dimethylhept-4-en-2-ol
Molekylvægt (g/mol) 469.75
Formel vægt 469.39