missing translation for 'onlineSavingsMsg'
Få mere at vide

25-Hydroxy Cholesterol, TRC

Artikelnummer. 30407020
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407020 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30407020 Leverandør Toronto Research Chemicals Leverandørnr. TRCH918030100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 25-Hydroxy Cholesterol
CAS 2140-46-7
Renhedsgrad noter HPLC
Molekylær formel C27 H46 O2
Mængde 100 mg
Synonym Cholest-5-ene-3,25-diol, (3β)-, Cholest-5-ene-3β,25-diol (8CI), (3β)-Cholest-5-ene-3,25-diol, 25-Hydroxy-5-cholestene-3β-ol, 25-Hydroxycholesterol, 5-Cholesten-3β,25-diol, 25-Hydroxy Cholesterol
InChI formel InChI=1S/C27H46O2/c1-18(7-6-14-25(2,3)29)22-10-11-23-21-9-8-19-17-20(28)12-15-26(19,4)24(21)13-16-27(22,23)5/h8,18,20-24,28-29H,6-7,9-17H2,1-5H3/t18-,20+,21+,22-,23+,24+,26+,27-/m1/s1
SMIL C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-17-[(2R)-6-hydroxy-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 402.65
Formel vægt 402.3498
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.