missing translation for 'onlineSavingsMsg'
Få mere at vide

24-Nor Ursodeoxycholic Acid, TRC

Artikelnummer. 30134332
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
10 mg
50 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134332 10 mg -
30133065 50 mg -
30003446 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30134332 Leverandør Toronto Research Chemicals Leverandørnr. TRCN86530010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99697-24-2
Molekylær formel C23 H38 O4
Molekylvægt (g/mol) 378.55
Synonym (3α,5β,7β)-3,7-Dihydroxy-24-norcholan-23-oic Acid;
IUPAC navn (3R)-3-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]butanoic acid
SMIL C[C@H](CC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Kemisk navn eller materiale 24-Nor Ursodeoxycholic Acid
Mængde 10 mg
InChI formel InChI=1S/C23H38O4/c1-13(10-20(26)27)16-4-5-17-21-18(7-9-23(16,17)3)22(2)8-6-15(24)11-14(22)12-19(21)25/h13-19,21,24-25H,4-12H2,1-3H3,(H,26,27)/t13-,14+,15-,16-,17+,18+,19+,21+,22+,23-/m1/s1
Formel vægt 378.277
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.