Få mere at vide
24-Nor Ursodeoxycholic Acid, TRC
Artikelnummer. 30133065
Skift visning
Mængde:
2.5 mg
10 mg
50 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133065 50 mg -
30134332 10 mg -
30003446 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133065 Leverandør Toronto Research Chemicals Leverandørnr. TRCN86530050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 99697-24-2
Molekylær formel C23 H38 O4
Molekylvægt (g/mol) 378.55
Synonym (3α,5β,7β)-3,7-Dihydroxy-24-norcholan-23-oic Acid;
IUPAC navn (3R)-3-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]butanoic acid
SMIL C[C@H](CC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 24-Nor Ursodeoxycholic Acid
Mængde 50 mg
InChI formel InChI=1S/C23H38O4/c1-13(10-20(26)27)16-4-5-17-21-18(7-9-23(16,17)3)22(2)8-6-15(24)11-14(22)12-19(21)25/h13-19,21,24-25H,4-12H2,1-3H3,(H,26,27)/t13-,14+,15-,16-,17+,18+,19+,21+,22+,23-/m1/s1
Formel vægt 378.277