Få mere at vide
24-Hydroxycholesterol, TRC
Artikelnummer. 30005192
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005192 100 mg
30012051 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005192 Leverandør Toronto Research Chemicals Leverandørnr. TRCH918040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 30271-38-6
Molekylær formel C27H46O2
Molekylvægt (g/mol) 402.65
Synonym Cholest-5-ene-3β,24-diol, (3β)-Cholest-5-ene-3,24-diol
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-17-[(2R)-5-hydroxy-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC(C)C(O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 24-Hydroxycholesterol
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25?,26+,27-/m1/s1
Formel vægt 402.3498
Procent renhed >95%