Learn More
Hamilton™ 23 gauge, Hat Needle
Hat type needles are designed to be connected directly to a flat bottom valve port.
Brand:  Hamilton™ 90823
Additional Details : Weight : 0.02400kg
Product Code. 11583224
  • 921.00 DKK / Pack of 3
Estimated Shipment: 03-04-2024
to see stock.
Description
Description
Specification Sheet
Gauge23 gauge
Point Style2, 3, or 4
Needle LengthCustom Needle Length
Sales Quantity3 needles per pack
HubHat Needle
Specifications
Specifications
Custom Length
0.375 to 12 in.
3/Pk.
23 G
Hat Needle
2, 3, or 4