missing translation for 'onlineSavingsMsg'
Learn More

(22S)-Budesonide, TRC

Product Code. 30133976
Change view
Click to view available options
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30133976 10 mg
30133107 25 mg
30007963 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133976 Supplier Toronto Research Chemicals Supplier No. TRCB68950510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51372-28-2
Molekylær formel C25H34O6
Molekylvægt (g/mol) 430.53
Synonym (11beta,16alpha)-16,17-[(1S)-Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide Epimer A (USP)
IUPAC navn (1S,2S,4R,6S,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
SMIL CCC[C@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Kemisk navn eller materiale (22S)-Budesonide
Mængde 10 mg
InChI formel InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21-,22+,23-,24-,25+/m0/s1
Formel vægt 430.2355
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.