Få mere at vide
(22E)-Ergosta-4,6,8(14),22-tetraen-3-one, TRC
Artikelnummer. 17917834
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17917834 10 mg
17939644 25 mg
17997624 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17917834 Leverandør Toronto Research Chemicals Leverandørnr. TRCE59917010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19254-69-4
Molekylær formel C28 H40 O
Molekylvægt (g/mol) 392.62
Synonym (22E)-4,6,8(14),22-Ergostatetraen-3-one, Ergone (22E,24R)-Ergosta-4,6,8(14),22-tetraen-3-one, Ergosta-4,6,8(14),22-tetraen-3-one
IUPAC navn (9R,10R,13R,17R)-10,13-dimethyl-17-[(E,1R,4R)-1,4,5-trimethylhex-2-enyl]-1,2,9,11,12,15,16,17-octahydrocyclopenta[a]phenanthren-3-one
SMIL CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CCC2=C3C=CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale (22E)-Ergosta-4,6,8(14),22-tetraen-3-one
Mængde 10 mg
InChI formel InChI=1S/C28H40O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,17-20,24,26H,11-16H2,1-6H3/b8-7+/t19-,20+,24+,26-,27-,28+/m0/s1
Formel vægt 392.3079