Få mere at vide
22,23-Didehydro Selamectin(>90%), TRC
Artikelnummer. 30369131
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369131 50 mg
30368965 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30369131 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43944550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 165108-09-8
Molekylær formel C43 H61 N O11
Molekylvægt (g/mol) 767.94
Synonym Avermectin A1a, 25-cyclohexyl-4'-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-5-(hydroxyimino)- (9CI), (5EZ)-22,23-Didehydroselamectin, (2aE,2'S,4E,5'S,6S,6'R,7S,8E,11R,15S,17aR,20EZ,20aR,20bS)-6'-cyclohexyl-7-[(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)oxy]-20b-hydroxy-20-(hydroxyimino)-5',6,8,19-tetramethyl-5',6,6',7,10,11,14,15,17a,20,20a,20b-dodecahydrospiro[2H,17H-11,15-methanofuro[4,3,2-pq][2,6]benzodioxacyclooctadecine-13,2'-pyran]-17-one, (5EZ,25R)-25-cyclohexyl-4'-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-5-(hydroxyimino)avermectin A1a
SMIL CO[C@H]1C[C@H](O[C@H]2[C@@H](C)\C=C\C=C\3/CO[C@@H]4C(=NO)C(=C[C@@H](C(=O)O[C@H]5C[C@@H](C\C=C\2/C)O[C@@]6(C5)O[C@H](C7CCCCC7)[C@@H](C)C=C6)[C@]34O)C)O[C@@H](C)[C@@H]1O
Tekniske data
Kemisk navn eller materiale (5EZ)-22,23-Didehydro Selamectin
Mængde 50 mg
InChI formel InChI=1S/C43H61NO11/c1-24-11-10-14-30-23-50-40-36(44-48)27(4)19-33(43(30,40)47)41(46)52-32-20-31(16-15-25(2)38(24)53-35-21-34(49-6)37(45)28(5)51-35)54-42(22-32)18-17-26(3)39(55-42)29-12-8-7-9-13-29/h10-11,14-15,17-19,24,26,28-29,31-35,37-40,45,47-48H,7-9,12-13,16,20-23H2,1-6H3/b11-10+,25-15+,30-14+,44-36?/t24-,26-,28-,31+,32-,33-,34-,35-,37-,38-,39-,40+,42+,43+/m0/s1
Formel vægt 767.4245