Learn More
Hamilton™ 21 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Brand:  Hamilton™ 22021-02
Additional Details : Weight : 0.01000kg
Product Code. 12341808
  • 746.00 DKK / Pack of 6
Estimated Shipment: 29-04-2024
to see stock.
Description
Description
Specification Sheet
Gauge21 gauge
Point Style5 or AS
Needle LengthCustom Needle Length
AutoclavableYes
Sales Quantity6 needles per pack
HubNo Hub
Specifications
Specifications
21 G
Hubless Needles