Få mere at vide
21-Deschloromethyl-21-O-Ethoxycarbonyl Loteprednol Etabonate, TRC
Artikelnummer. 30375050
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375050 250 mg
30371569 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30375050 Leverandør Toronto Research Chemicals Leverandørnr. TRCE895120250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 133991-62-5
Molekylær formel C26H34O9
Molekylvægt (g/mol) 490.54
Synonym (11beta,17alpha)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid Anhydride with Ethyl Hydrogen Carbonate, Prednisolone Dicarbonate
SMIL CCOC(=O)OC(=O)[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)OC(=O)OCC
Tekniske data
Kemisk navn eller materiale 21-Deschloromethyl-21-O-Ethoxycarbonyl Loteprednol Etabonate
Mængde 250 mg
InChI formel InChI=1S/C26H34O9/c1-5-32-22(30)34-21(29)26(35-23(31)33-6-2)12-10-18-17-8-7-15-13-16(27)9-11-24(15,3)20(17)19(28)14-25(18,26)4/h9,11,13,17-20,28H,5-8,10,12,14H2,1-4H3/t17-,18-,19-,20+,24-,25-,26-/m0/s1
Formel vægt 490.2203