Få mere at vide
21-Desacetoxydeflazacort, Mikromol™
Artikelnummer. 30051493
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051493 25 mg
30050132 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051493 Leverandør Mikromol Leverandørnr. MM1224.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
21-Desacetoxydeflazacort
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 21-Deacetoxydeflazacort
CAS 13649-88-2
Analyt- eller komponentnavne 21-Deacetoxydeflazacort
Forsendelsestilstand Room Temperature
Molekylær formel C23 H29 N O4
Certificeringer/compliance ISO 17025
Synonym 21-Desacetoxydeflazacort, (11beta,16beta)-11-Hydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 5'betaH-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11beta-hydroxy-2'-methyl- (8CI)
InChI formel InChI=1S/C23H29NO4/c1-12(25)23-19(28-13(2)24-23)10-17-16-6-5-14-9-15(26)7-8-21(14,3)20(16)18(27)11-22(17,23)4/h7-9,16-20,27H,5-6,10-11H2,1-4H3/t16-,17-,18-,19+,20+,21-,22-,23+/m0/s1
SMIL CC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C
Molekylvægt (g/mol) 383.48
Mængde 25 mg
Formel vægt 383.2097 g/mol