Få mere at vide
21-Deacetoxy Deflazacort, TRC
Artikelnummer. 30359717
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359717 10 mg
30359679 5 mg
30352458 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30359717 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19800010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13649-88-2
Molekylær formel C23 H29 N O4
Molekylvægt (g/mol) 383.48
Synonym (11β,16β)-11-Hydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 5'βH-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11β-hydroxy-2'-methyl- (8CI)
IUPAC navn (1S,2S,4R,8S,9S,11S,12S,13R)-8-acetyl-11-hydroxy-6,9,13-trimethyl-5-oxa-7-azapentacyclo[10.8.0.02,9.04,8.013,18]icosa-6,14,17-trien-16-one
SMIL CC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C
Tekniske data
Kemisk navn eller materiale 21-Deacetoxydeflazacort
Mængde 10 mg
InChI formel InChI=1S/C23H29NO4/c1-12(25)23-19(28-13(2)24-23)10-17-16-6-5-14-9-15(26)7-8-21(14,3)20(16)18(27)11-22(17,23)4/h7-9,16-20,27H,5-6,10-11H2,1-4H3/t16-,17-,18-,19+,20+,21-,22-,23+/m0/s1
Formel vægt 383.2097