Få mere at vide
21-Acetoxy-11Beta-hydroxy-16Alpha,17Alpha-propylmethylenedioxpregna-1,4-diene-3,20-dione, TRC
Artikelnummer. 17992815
Skift visning
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17992815 1 mg
17910595 100 mg
17920375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17992815 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1657401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51333-05-2
Molekylær formel C27 H36 O7
Molekylvægt (g/mol) 472.57
Synonym (11β,16α)-21-(Acetyloxy)-16,17-[butylidenebis(oxy)]-11-hydroxy-pregna-1,4-diene-3,20-dione, 21-(Acetoxy)-16α,17α-(butylidenedioxy)-11β-hydroxypregna-1,4-diene-3,20-dione, 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate, Budesonide Imp. K (EP)
IUPAC navn [2-[(1S,2S,4R,8S,9S,11S,12S,13R)-11-hydroxy-9,13-dimethyl-16-oxo-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
SMIL CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COC(=O)C
Tekniske data
Kemisk navn eller materiale 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate
Analyt- eller komponentnavne 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C27H36O7/c1-5-6-23-33-22-12-19-18-8-7-16-11-17(29)9-10-25(16,3)24(18)20(30)13-26(19,4)27(22,34-23)21(31)14-32-15(2)28/h9-11,18-20,22-24,30H,5-8,12-14H2,1-4H3/t18-,19-,20-,22+,23?,24+,25-,26-,27+/m0/s1
Løsningstype Neat
Industry Type Forensics
Mængde 1 mg
Formel vægt 472.2461
Procent renhed >95%