missing translation for 'onlineSavingsMsg'
Få mere at vide

21-Acetoxy-11Beta-hydroxy-16Alpha,17Alpha-propylmethylenedioxpregna-1,4-diene-3,20-dione, TRC

Product Code. 17910595
Change view
Click to view available options
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17910595 100 mg
17920375 10 mg
17992815 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17910595 Supplier Toronto Research Chemicals Supplier No. TRCA165740100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51333-05-2
Molekylær formel C27 H36 O7
Molekylvægt (g/mol) 472.57
Synonym (11β,16α)-21-(Acetyloxy)-16,17-[butylidenebis(oxy)]-11-hydroxy-pregna-1,4-diene-3,20-dione, 21-(Acetoxy)-16α,17α-(butylidenedioxy)-11β-hydroxypregna-1,4-diene-3,20-dione, 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate, Budesonide Imp. K (EP)
IUPAC navn [2-[(1S,2S,4R,8S,9S,11S,12S,13R)-11-hydroxy-9,13-dimethyl-16-oxo-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
SMIL CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COC(=O)C

Specifications

Kemisk navn eller materiale 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate
Analyt- eller komponentnavne 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C27H36O7/c1-5-6-23-33-22-12-19-18-8-7-16-11-17(29)9-10-25(16,3)24(18)20(30)13-26(19,4)27(22,34-23)21(31)14-32-15(2)28/h9-11,18-20,22-24,30H,5-8,12-14H2,1-4H3/t18-,19-,20-,22+,23?,24+,25-,26-,27+/m0/s1
Løsningstype Neat
Industry Type Forensics
Mængde 100 mg
Formel vægt 472.2461
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.