missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(20S)-21-Hydroxy-20-methylpregn-4-en-3-one, TRC
Specifications
Specifications
| Kemisk navn eller materiale | (20S)-21-Hydroxy-20-methylpregn-4-en-3-one |
| Mængde | 100 mg |
| InChI formel | InChI=1S/C22H34O2/c1-14(13-23)18-6-7-19-17-5-4-15-12-16(24)8-10-21(15,2)20(17)9-11-22(18,19)3/h12,14,17-20,23H,4-11,13H2,1-3H3/t14-,17+,18-,19+,20+,21+,22-/m1/s1 |
| Formel vægt | 330.2559 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?