missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ 20 mM Sodium Acetate

Artikelnummer. 30286137
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 L
20 L
50 L
100 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30286137 5 L
30286136 20 L
30286139 50 L
30286132 100 L
30286135 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30286137 Leverandør Gibco™ Leverandørnr. A59148BA

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.

Features

  • Available in a range of pack sizes
  • Established shelf life; set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
TRUSTED_SUSTAINABILITY

Tekniske data

Form Clear, colorless solution
Produkttype Sodium Acetate
Sterilitet Sterile-filtered
Steriliseringsmetode Sterile-filtered
Formulering Acetate Buffer
Fremstillingskvalitet CGMP
Mængde 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.