Learn More
Fisherbrand™ 20mm Crimp Seal, Silver, Aluminum Cap, 10mm Center Hole, Assembled Septum
Product Code. 11778246 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Beige, White
Blue, White
Dark Gray
Red-Orange, Transparent
Silver, Blue Transparent, White
Silver, Blue, Gray
Silver, Gray, Red
Silver, White
Material (Liner):
Butyl
Natural Rubber/TEF
PTFE/Butyl
Silicone/Aluminum
Silicone/PTFE
Thickness (Metric):
1.3 mm
1.5 mm
3 mm
3.2 mm
Unit Size:
Pack of 1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material (Liner) Thickness (Metric) Unit Size
11778246 Silver, Blue, Gray PTFE/Butyl 3 mm Pack of 1000
11742458 Silver, Gray, Red PTFE/Butyl 3 mm Pack of 1000
11891653 Silver, White Silicone/Aluminum 3 mm Pack of 1000
11590505 Beige, White Silicone/PTFE 3.2 mm Pack of 1000
11793127 Silver, Blue Transparent, White Silicone/PTFE 3 mm Pack of 1000
11845030 Dark Gray Butyl 3 mm Pack of 1000
11884011 Blue, White Silicone/PTFE 1.5 mm Pack of 1000
12616775 Red-Orange, Transparent Natural Rubber/TEF 1.3 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11778246
Discount Lab Supplies
Use promo code 28170
View all promotions
Forventet forsendelse: 14-12-2026
Add to basket
Add to basket
Pre-assembled caps and septa are convenient and minimize contamination from handling.
  • 20mm
  • Combination Seal
  • aluminum
  • Cap
  • li>Center hole
TRUSTED_SUSTAINABILITY
Specifications
Description Pharma-Fix-Septa
Material (Liner) PTFE/Butyl
Color Silver, Blue, Gray
Diameter (Metric) Outer 20 mm
Material Aluminum
For Use With (Application) Headspace
Quantity 1000
Thickness (Metric) 3 mm
Closure Size 20 mm
For Use With (Equipment) 20 mm Crimp Top Vials
Green Features ACT Ecolabel (Silver)
Product Type Crimp Cap