Learn More
Fisher Chemical 200LT HEXANES, FOR ANALYSIS, 95% N-HEXANE APPROX
Brand: Fisher Chemical H/0355/27
9415.73 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
200LT HEXANES, FOR ANALYSIS, 95% N-HEXANE APPROX
Description
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2539324 | Certificate of Analysis | 09/06/2025 | H/0355/25,H/0355/15,H/0355/27,H/0355/17,H/0355/21,H/0355/17X,H/0355/17Z |
2534196 | Certificate of Analysis | 09/06/2025 | H/0355/25,H/0355/15,H/0355/27,H/0355/17,H/0355/21,H/0355/17X,H/0355/17Z |
2534196 | Certificate of Analysis | 02/06/2025 | H/0355/25,H/0355/27,H/0355/21,H/0355/17X,H/0355/17Z |
2532504 | Certificate of Analysis | 24/05/2025 | H/0355/25,H/0355/15,H/0355/27,H/0355/17,H/0355/21,H/0355/17X,H/0355/17Z |
2532504 | Certificate of Analysis | 17/05/2025 | H/0355/25,H/0355/27,H/0355/21,H/0355/17X,H/0355/17Z |