missing translation for 'onlineSavingsMsg'
Learn More
Learn More
20(R)-Ginsenoside Rh2, MedChemExpress

Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
It has anticancer effects via blocking cell proliferation and causing G1 phase arrest. 20(R)-Ginsenoside Rh2 induces apoptosis, and has anti-inflammatory and antioxidative activity. 20(R)-Ginsenoside Rh2 inhibits the replication and proliferation of mouse and human gammaherpesvirus 68 (MHV-68) with an IC50 of 2.77 ÎĽM for murine MHV-68.
Specifications
Specifications
Chemical Name or Material | 20(R)-Ginsenoside Rh2 |
Physical Form | Solid |
CAS | 112246-15-8 |
Color | White |
Quantity | 10 mg |
Purity Grade Notes | Research |
Molecular Formula | C36H62O8 |
For Use With (Application) | Cancer-Kinase/protease |
Solubility Information | DMSO : 125 mg/mL (200.68 mM; Need ultrasonic) |
SMILES | O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1O[C@H]2CC[C@]3(C)[C@@]4([H])C[C@@H](O)[C@@H]5[C@@H]([C@](C)(O)CC/C=C(C)\C)CC[C@](C)5[C@@](C)4CC[C@@]3([H])C2(C)C |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
20(R)-Ginsenoside Rh2, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction