Få mere at vide
20-Dihydro-corticosterone 21-Carboxylic Acid, TRC
Artikelnummer. 30369689
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369689 100 mg
30379959 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30369689 Leverandør Toronto Research Chemicals Leverandørnr. TRCD452405100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75879-79-7
Molekylær formel C21 H30 O5
Molekylvægt (g/mol) 362.46
Synonym Pregn-4-en-21-oic acid, 11,20-dihydroxy-3-oxo-, (11β)-, (11β)-11,20-Dihydroxy-3-oxopregn-4-en-21-oic acid, 11β,20-Dihydroxy-3-oxopregn-4-en-21-oic acid, 20-Dihydrocorticosterone-21-carboxylic acid
IUPAC navn 2-hydroxy-2-[(8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]acetic acid
SMIL C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(O)C(=O)O
Tekniske data
Kemisk navn eller materiale 20-Dihydro-corticosterone 21-Carboxylic Acid
Mængde 100 mg
InChI formel InChI=1S/C21H30O5/c1-20-8-7-12(22)9-11(20)3-4-13-14-5-6-15(18(24)19(25)26)21(14,2)10-16(23)17(13)20/h9,13-18,23-24H,3-8,10H2,1-2H3,(H,25,26)/t13-,14-,15+,16-,17+,18?,20-,21-/m0/s1
Formel vægt 362.2093