Få mere at vide
20-Deoxyingenol, MedChemExpress
Mængde:
5 mg
10 mg
Description
20-Deoxyingenol, a diterpene, is isolated from the roots of Euphorbia kansui. 20-Deoxyingenol can promote autophagy and lysosomal biogenesis by promoting the nuclear translocation of transcription factor EB (TFEB) in vitro. 20-Deoxyingenol can be used for the research of osteoarthritis (OA).
Specifications
Specifications
| Kemisk navn eller materiale | 20-Deoxyingenol |
| CAS | 54706-99-9 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C20H28O4 |
| Mængde | 5 mg |
| Opløselighedsinformation | DMSO : 50 mg/mL (150.41 mM; Need ultrasonic) |
| SMIL | O=C1[C@@]2(C=C(C)[C@@H]3O)[C@]3(O)[C@H](O)C(C)=C[C@@]1([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C |
| Molekylvægt (g/mol) | 332.43 |
| Formel vægt | 332.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.