Learn More
Ohaus™ 2 x Centrifuge Buckets, Frontier™ R-S4x100/5MIS Swing Out Rotor
Carrier buckets for centrifuge tubes, suitable for the R-S4x100/5MIS Swing Out Rotor.
Brand: Ohaus™ 30314881
2178.69 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Swinging bucket holds specifically sized tubes for centrifugation
- Suitable for use in Frontier™ R-S4x100/5MIS Swing Out Rotors
Specifications
Bucket, 2x50ml D29mm FA, w/ Rack, 2/pk | |
2 x 50 mL | |
R-S4x100/5MIS Swing Out Rotor | |
2 |
Fixed Angle Rotor | |
29 mm | |
2 Buckets |
Product Content Correction
Product Title
Ohaus™ 2 x Centrifuge Buckets, Frontier™ R-S4x100/5MIS Swing Out Rotor > 2 x 50 mL
Spot an opportunity for improvement?Share a Content Correction