Learn More
Alfa Aesar™ 2-(Trimethylsilyl)thiophene, 97%
Brand: Alfa Aesar™ H61170.06
Additional Details : CAS Number : 18245-28-8 Weight : 0.00500kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Quantity | 5g |
---|
Description
Applications2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.
Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Chemical Identifiers
18245-28-8 | |
156.318 | |
OANGLGSZPSFVDY-UHFFFAOYSA-N | |
140360 | |
C[Si](C)(C)C1=CC=CS1 |
C7H12SSi | |
MFCD00005415 | |
2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane | |
trimethyl(thiophen-2-yl)silane |
Specifications
2-(Trimethylsilyl)thiophene | |
41°C (106°F) | |
5g | |
C7H12SSi | |
UN1993 | |
OANGLGSZPSFVDY-UHFFFAOYSA-N | |
trimethyl(thiophen-2-yl)silane | |
140360 | |
97% |
0.926 | |
1.497 | |
18245-28-8 | |
MFCD00005415 | |
2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane | |
C[Si](C)(C)C1=CC=CS1 | |
156.318 | |
156.32 |