Learn More
2-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific™
Code produit 11379945
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
Conditionnement:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
11379945 5 g 5g
11369945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11379945 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur L19913.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13333-97-6
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00068896
InChI Key XVGQHRKNXSUPEF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol
PubChem CID 2777889
IUPAC Name 2-(trifluoromethyl)benzenethiol
SMILES C1=CC=C(C(=C1)C(F)(F)F)S
Specifications
Density 1.35
Boiling Point 175°C to 177°C
Flash Point 68°C (154°F)
Refractive Index 1.499
Quantity 5 g
UN Number UN2810
Beilstein 2441965
Sensitivity Air Sensitive
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)thiophenol