missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Trifluoromethyl)pyridine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63315.03

Additional Details : CAS Number : 131747-41-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469948

  • 1431.61 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Description

Description

Applications
2-(Trifluoromethyl)pyridine-4-carboxylic acid, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

131747-41-6
191.109
BZFGKBQHQJVAHS-UHFFFAOYSA-N
14761447
C1=CN=C(C=C1C(=O)O)C(F)(F)F
C7H4F3NO2
MFCD07774132
2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl
2-(trifluoromethyl)pyridine-4-carboxylic acid

Specifications

2-(Trifluoromethyl)pyridine-4-carboxylic acid
217°C to 223°C
131747-41-6
MFCD07774132
BZFGKBQHQJVAHS-UHFFFAOYSA-N
2-(trifluoromethyl)pyridine-4-carboxylic acid
14761447
Solid
White
1g
C7H4F3NO2
2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl
C1=CN=C(C=C1C(=O)O)C(F)(F)F
191.109
191.11
97%
Videos
SDS