Learn More
2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3107-34-4 |
| Molecular Formula | C7H8ClF3N2 |
| MDL Number | MFCD00102619 |
| Quantity | 100 g |
| SMILES | NNC1=CC=CC=C1C(F)(F)F.[H]Cl |
| Molecular Weight (g/mol) | 212.60 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(Trifluoromethyl)phenylhydrazine hydrochloride |