missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Catalog Number 11497137
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
447-60-9
C8H4F3N
171.122
MFCD00001791
SOZGHDCEWOLLHV-UHFFFAOYSA-N
2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
67972
2-(trifluoromethyl)benzonitrile
C1=CC=C(C(=C1)C#N)C(F)(F)F
This item is not returnable. View return policy
447-60-9
C8H4F3N
171.122
MFCD00001791
SOZGHDCEWOLLHV-UHFFFAOYSA-N
2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
67972
2-(trifluoromethyl)benzonitrile
C1=CC=C(C(=C1)C#N)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17°C to 18°C
Density 1.294
Boiling Point 204°C to 206°C
Flash Point 90°C (194°F)
Refractive Index 1.4632
Quantity 50 g
UN Number UN3276
Beilstein 2048152
Formula Weight 171.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'