Få mere at vide
2-Tricosanone, TRC
Artikelnummer. 30451582
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451582 1 g
30456754 100 mg
30456522 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30451582 Leverandør Toronto Research Chemicals Leverandørnr. TRCT9020801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7320-54-9
Molekylær formel C23H46O
Molekylvægt (g/mol) 338.61
IUPAC navn tricosan-2-one
SMIL CCCCCCCCCCCCCCCCCCCCCC(=O)C
Tekniske data
Kemisk navn eller materiale 2-Tricosanone
Mængde 1 g
InChI formel InChI=1S/C23H46O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(2)24/h3-22H2,1-2H3
Formel vægt 338.3549