Learn More
2-(Trichloroacetyl)pyrrole, TRC
Product Code. 30454126
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30454126 5 g 5g
1 options
Product Code. 30454126 Supplier Toronto Research Chemicals Supplier No. TRCT7740855G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Trichloroacetyl)pyrrole
CAS 35302-72-8
Molecular Formula C6H4Cl3NO
Quantity 5 g
Synonym 2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone, 2-(Trichloroacetyl)-1H-pyrrole, NSC 272669, Pyrrol-2-yl Trichloromethyl Ketone
InChI Formula InChI=1S/C6H4Cl3NO/c7-6(8,9)5(11)4-2-1-3-10-4/h1-3,10H
SMILES ClC(Cl)(Cl)C(=O)c1ccc[nH]1
IUPAC Name 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
Molecular Weight (g/mol) 212.46
Formula Weight 210.9358