missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Thioxothiazolidine-4-carboxylic Acid, TRC

Product Code. 30456070
Change view
Click to view available options
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30456070 250 mg
30450117 50 mg
30451123 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456070 Supplier Toronto Research Chemicals Supplier No. TRCT368830250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20933-67-9
Molekylær formel C4 H5 N O2 S2
Molekylvægt (g/mol) 163.22
Synonym 4-Thiazolidinecarboxylic acid, 2-thioxo-, 2-Thioxo-4-thiazolidinecarboxylic acid, TTCA, TTCA (thiram metabolite), Thiazolidine-2-thione-4-carboxylic acid
IUPAC navn 2-sulfanylidene-1,3-thiazolidine-4-carboxylic acid
SMIL OC(=O)C1CSC(=S)N1

Specifications

Kemisk navn eller materiale 2-Thioxothiazolidine-4-carboxylic Acid
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)
Formel vægt 162.9762
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.