missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals

Catalog Number p-199738
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
27757-85-3
C5H7NS
113.18
MFCD00005460
FKKJJPMGAWGYPN-UHFFFAOYSA-N
2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
34005
thiophen-2-ylmethanamine
C1=CSC(=C1)CN
This item is not returnable. View return policy
27757-85-3
C5H7NS
113.18
MFCD00005460
FKKJJPMGAWGYPN-UHFFFAOYSA-N
2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
34005
thiophen-2-ylmethanamine
C1=CSC(=C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 95.0°C to 99.0°C (28.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5650 to 1.5680
Quantity 25 g
Beilstein 18, IV, 7096
Specific Gravity 1.1
Formula Weight 113.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethylamine, 90-96%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenemethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'