Få mere at vide
2-Thiocytidine, TRC
Artikelnummer. 30451293
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451293 25 mg
30451949 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451293 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34441825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13239-97-9
Molekylær formel C9H13N3O4S
Molekylvægt (g/mol) 259.28
IUPAC navn 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2-thione
SMIL NC1=NC(=S)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O
Tekniske data
Kemisk navn eller materiale 2-Thiocytidine
Mængde 25 mg
InChI formel InChI=1S/C9H13N3O4S/c10-5-1-2-12(9(17)11-5)8-7(15)6(14)4(3-13)16-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,17)/t4-,6-,7-,8-/m1/s1
Formel vægt 259.0627