Få mere at vide
2-tert-Butyldimethylsilyloxyethanol-d4, TRC
Artikelnummer. 30342832
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30342832 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342832 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69147210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-tert-Butyldimethylsilyloxyethanol-d4
CAS 764650-43-3
Molekylær formel C8H16D4O2Si
Mængde 10 mg
InChI formel InChI=1S/C8H20O2Si/c1-8(2,3)11(4,5)10-7-6-9/h9H,6-7H2,1-5H3/i6D2,7D2
SMIL [2H]C([2H])(O)C([2H])([2H])O[Si](C)(C)C(C)(C)C
IUPAC navn 2-[tert-butyl(dimethyl)silyl]oxy-1,1,2,2-tetradeuterioethanol
Molekylvægt (g/mol) 180.35