Få mere at vide
2-tert-Butyl-5-methyl-1,3-dioxane-5-carboxylic Acid Vinyl Ester-d3, TRC
Artikelnummer. 30333907
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30333907 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333907 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69330210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-tert-Butyl-5-methyl-1,3-dioxane-5-carboxylic Acid Vinyl Es-D3
CAS 1189464-68-3
Molekylær formel C4 H5 N3 S
Mængde 10 mg
InChI formel InChI=1S/C4H5N3S/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)
SMIL NC1=CC=NC(=S)N1
IUPAC navn 6-amino-1H-pyrimidine-2-thione
Molekylvægt (g/mol) 127.168
Formel vægt 127.02