Få mere at vide
2-tert-Butyl-4-hydroxyanisole-d3, TRC
Artikelnummer. 30343321
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343321 5 mg
30333359 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343321 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6993025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-tert-Butyl-4-hydroxyanisole-d3
CAS 1794892-02-6
Molekylær formel C11 D3 H13 O2
Mængde 5 mg
Synonym 3-(1,1-Dimethylethyl)-4-methoxyphenol, 2-(1,1-Dimethylethyl)-4-hydroxyanisole, 2-tert-Butyl-4-hydroxyanisole, 2-tert-Butyl-p-hydroxyanisole, 3-tert-Butyl-4-methoxyphenol, 4-Methoxy-3-(1,1-dimethylethyl)phenol, 4-Methoxy-3-tert-butylphenol, tert-Butyl-3-methoxy-4-phenol
InChI formel InChI=1S/C11H16O2/c1-11(2,3)9-7-8(12)5-6-10(9)13-4/h5-7,12H,1-4H3/i4D3
SMIL [2H]C([2H])([2H])Oc1ccc(O)cc1C(C)(C)C
IUPAC navn 3-tert-butyl-4-(trideuteriomethoxy)phenol
Molekylvægt (g/mol) 183.262
Formel vægt 183.134