Få mere at vide
2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid (alpha-Cyclopentylmandelic Acid), Mikromol™
Beskrivelse
2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid (alpha-Cyclopentylmandelic Acid)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid |
| CAS | 427-49-6 |
| Analyt- eller komponentnavne | 2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13H16O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | alpha-Cyclopentylmandelic Acid, Glycopyrronium Bromide Imp. J (EP), Glycopyrrolate USP Related Compound C, Glycopyrrolate USP RC C, 2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid, Glycopyrrolate Related Compound C, Glycopyrronium Bromide Impurity J, Glycopyrronium Impurity J |
| InChI formel | InChI=1S/C13H16O3/c14-12(15)13(16,11-8-4-5-9-11)10-6-2-1-3-7-10/h1-3,6-7,11,16H,4-5,8-9H2,(H,14,15) |
| SMIL | OC(=O)C(O)(C1CCCC1)c2ccccc2 |
| IUPAC navn | 2-cyclopentyl-2-hydroxy-2-phenyl-acetic acid |
| Vis mere |