Få mere at vide
2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride) |
| CAS | 4024-34-4 |
| Molekylær formel | C18 H23 N O . Cl H |
| Mængde | 10 mg |
| Synonym | 4-Methyldiphenhydramine Hydrochloride, (RS)-N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamine Hydrochloride, N,N-Dimethyl-2-[(RS)-(4-methylphenyl)(phenyl)methoxy]ethanamine Hydrochloride, 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. F (EP) as Hydrochloride, para-Methylbenzyl Isomer Hydrochloride of Orphenadrine, Dimenhydrinate Imp. G (EP) as Hydrochloride, Diphenhydramine Hydrochloride Imp. B (EP) as Hydrochloride |
| InChI formel | InChI=1S/C18H23NO.ClH/c1-15-9-11-17(12-10-15)18(20-14-13-19(2)3)16-7-5-4-6-8-16;/h4-12,18H,13-14H2,1-3H3;1H |
| SMIL | Cl.CN(C)CCOC(c1ccccc1)c2ccc(C)cc2 |
| IUPAC navn | N,N-dimethyl-2-[(4-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride |
| Molekylvægt (g/mol) | 305.84 |
| Formel vægt | 305.1546 |